2-Phenoxynicotinonitrile ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A15376.14EA 2960 DKK
A15376.14 A15376.06
2-Phenoxynicotinonitrile ≥98%
2-Phenoxynicotinonitrile
Formel: C₁₂H₈N₂O
Molekylvægt: 196,21
Smeltepunkt: 107…109 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015473
CAS-nummer: 14178-15-5
EINECS: 238-035-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR