3-Amino-4-methylbenzamide 97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B25367.14EA 1630 DKK
B25367.14 B25367.06
3-Amino-4-methylbenzamide 97%
3-Amino-4-methylbenzamide
Formel: C₈H₁₀N₂O
Smeltepunkt: 127…131 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00035936
CAS-nummer: 19406-86-1
EINECS: 243-039-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR