1,2,4,5-Tetrakis(tert-butylthio)benzene ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAT2217-1GEA
1150
DKK
TCIAT2217-1G
1,2,4,5-Tetrakis(tert-butylthio)benzene ≥98.0% (by GC)
1,2,4,5-Tetrakis(tert-butylthio)benzene
Formel:
C₂₂H₃₈S₄ Molekylvægt: 430,8000 g/mol |
MDL Number:
MFCD08276431 CAS-nummer: 447463-65-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...