4-Bromo-2-chloro-5-methoxyphenylboronic acid ≥97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H53058.MDEA 746 DKK
H53058.MD H53058.03
4-Bromo-2-chloro-5-methoxyphenylboronic acid ≥97%
4-Bromo-2-chloro-5-methoxyphenylboronic acid
Formel: C₇H₇BBrClO₃
Molekylvægt: 265,30 g/mol
Smeltepunkt: 260…268 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06659862
CAS-nummer: 850567-94-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR