Ethyl-2-(methylthio)isobutyrate ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B22597.88EA 317 DKK
B22597.88 B22597.09
Ethyl-2-(methylthio)isobutyrate ≥98%
Ethyl-2-(methylthio)isobutyrate
Formel: C₇H₁₄O₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00053099
CAS-nummer: 49773-24-2
EINECS: 256-483-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR