1,3-Propanedisulphonic acid 50 - 60% in water
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAP1204-10GEA
493
DKK
TCIAP1204-10G
1,3-Propanedisulphonic acid 50 - 60% in water
1,3-Propanedisulphonic acid
Formel:
C₃H₈O₆S₂ Molekylvægt: 204,21 g/mol |
MDL Number:
MFCD00191482 CAS-nummer: 21668-77-9 UN nummer: 3265 ADR: 8,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Concentration (Neutralization titration) | 50.0 to 60.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...