4-Methyl-2-(methylthio)pyrimidine ≥98%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H28795.06EA
553
DKK
H28795.06
4-Methyl-2-(methylthio)pyrimidine ≥98%
4-Methyl-2-(methylthio)pyrimidine
Formel:
C₆H₈N₂S Molekylvægt: 140,21 g/mol Massefylde: 1,01 Flammepunkt: 104 °C (219 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00023242 CAS-nummer: 14001-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...