Methyl diphenylphosphinite ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD2557-5GEA 723 DKK
TCIAD2557-5G
Methyl diphenylphosphinite ≥98.0% (by GC)
Methyl diphenylphosphinite
Formel: C₁₃H₁₃OP
Molekylvægt: 216,22 g/mol
MDL Number: MFCD00048025
CAS-nummer: 4020-99-9
EINECS: 223-683-9
https://dk.vwr.com/stibo/web/std.lang.all/95/28/28979528.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR