4-(Difluoromethoxy)cinnamic acid ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H32170.MDEA 574 DKK
H32170.MD H32170.03
4-(Difluoromethoxy)cinnamic acid ≥97%
4-(Difluoromethoxy)cinnamic acid
Formel: C₁₀H₈F₂O₃
Smeltepunkt: 186…189 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01222822
CAS-nummer: 103962-07-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR