1-(3-Chlorophenyl)piperazine monohydrochloride

Leverandør: CERILLIANT CORPORATION
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

CERINMID907EA 0 DKK
CERINMID907
1-(3-Chlorophenyl)piperazine monohydrochloride
1-(3-Chlorophenyl)piperazine monohydrochloride
Formel: C₁₀H₁₄Cl₂N₂
Molekylvægt: 233,14 g/mol
MDL Number: MFCD00012764
CAS-nummer: 13078-15-4
EINECS: 235-976-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR