Graphite ink for tantalum capacitors
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
42193.30EA 1550 DKK
42193.30
Graphite ink for tantalum capacitors
Graphite ink
A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors