Graphite ink for tantalum capacitors
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
42193.30EA
1550
DKK
42193.30
Graphite ink for tantalum capacitors
Graphite ink
A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors