Pilocarpine nitrate

Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PROOBP503EA 3420 DKK
PROOBP503
Pilocarpine nitrate
Pilocarpine nitrate
Formel: C₁₁H₁₇N₃O₅
MDL Number: MFCD00078497
CAS-nummer: 148-72-1
EINECS: 205-723-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR