2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H54213.03EA 4210 DKK
H54213.03 H54213.MD
2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid ≥97%
2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid
Formel: C₁₁H₁₃BFNO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11504847
CAS-nummer: 1072951-41-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR