1-(Phenylethynyl)cyclohexanol ≥99%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B21808.06EA
1640
DKK
B21808.06
1-(Phenylethynyl)cyclohexanol ≥99%
1-(Phenylethynyl)cyclohexanol
Formel:
C₁₄H₁₆O Molekylvægt: 200,28 g/mol Smeltepunkt: 60…62 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00060883 CAS-nummer: 20109-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...