S-Furfuryl thiopropionate ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAF0578-25GEA
419
DKK
TCIAF0578-25G
S-Furfuryl thiopropionate ≥98.0% (by GC)
S-Furfuryl thiopropionate
Formel:
C₈H₁₀O₂S Molekylvægt: 170,23 g/mol Flammepunkt: 98°C |
MDL Number:
MFCD00040264 CAS-nummer: 59020-85-8 EINECS: 261-562-2 |
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...