Bis(cyclopentadienyl)dimethylhafnium ≥97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H26962.03EA
1580
DKK
H26962.03
Bis(cyclopentadienyl)dimethylhafnium ≥97%
Bis(cyclopentadienyl)dimethylhafnium
Formel:
C₁₂H₁₆Hf Molekylvægt: 338,745 g/mol Smeltepunkt: ∼ 118 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02093015 CAS-nummer: 37260-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...