2-Benzyloxy-5-fluorophenylboronic acid pinacol ester 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H62613.03EA 7060 DKK
H62613.03
2-Benzyloxy-5-fluorophenylboronic acid pinacol ester 96%
2-Benzyloxy-5-fluorophenylboronic acid pinacol ester
Formel: C₁₉H₂₂BFO₃
Molekylvægt: 328,19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22494088
CAS-nummer: 779331-48-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR