1,3-Dipropyl-8-phenylxanthine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J63554.MFEA 7550 DKK
J63554.MF
1,3-Dipropyl-8-phenylxanthine
1,3-Dipropyl-8-phenylxanthine

Selective A1 adenosine antagonist

Formel: C₁₅H₁₆N₄O₂
Molekylvægt: 312,37 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00209811
CAS-nummer: 85872-53-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR