1,3-Dibutylxanthine ≥98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B20491.36EA
5020
DKK
B20491.36
1,3-Dibutylxanthine ≥98%
1,3-Dibutylxanthine
Formel:
C₁₃H₂₀N₄O₂ Molekylvægt: 264,33 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00798591 CAS-nummer: 2850-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...