(Pentamethylcyclopentadienyl)tantalum tetrachloride
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
39631.03EA
2300
DKK
39631.03
39631.06
(Pentamethylcyclopentadienyl)tantalum tetrachloride
(Pentamethylcyclopentadienyl)tantalum tetrachloride
Formel:
C₁₀H₁₅Cl₄Ta Smeltepunkt: 220 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00070463 CAS-nummer: 71414-47-6 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...