5-Chloro-2-(methylamino)benzophenone

Leverandør: U.S. Pharmacopeia
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USPH1644319EA 7680 DKK
USPH1644319
5-Chloro-2-(methylamino)benzophenone
5-Chloro-2-(methylamino)benzophenone
Formel: C₁₄H₁₂ClNO
Molekylvægt: 245,71
MDL Number: MFCD00008284
CAS-nummer: 1022-13-5
EINECS: 213-822-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR