VWR®, LED Illuminated Table Magnifier
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
630-1959EA 285 DKK
630-1959 630-1957 630-1956 630-1954 630-1960
VWR®, LED Illuminated Table Magnifier
Forstørrelsesglas og lupper
These illuminated desktop magnifiers, gooseneck or articulated arm type, include highly polished glass.
  • Large viewing area
  • Cover protects lens from dust and scratches
  • Articulated arm models provide 200° head rotation
Suitable for biological and clinical laboratories, electronics and chemical industries.
Electrical data: 200 mA, 100 to 230 V, 50 to 60 Hz, 3,5 W (60 LED magnifiers); 200 mA, 100 to 230 V, 50 to 60 Hz, 2,5 W (40 LED magnifiers)
For information or technical assistance, please contact your local Avantor representative.
Advarsel: Only suitable for indoor use. When the lamp is replaced the cover must be fixed. Lamp must not be operated without the cover in place. Never leave the lens in direct sunlight or where light can shine through it, as this may cause a fire risk. Any damage to the external cable or lamp must be repaired or replaced by the manufacturer or a qualified person.