VWR®, LED illuminated bench magnifiers
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
630-1948EA 993 DKK
630-1948 630-1947 630-1951 630-1950
VWR®, LED illuminated bench magnifiers
Forstørrelsesglas og lupper
These illuminated magnifiers can be fixed on a table or bench.
  • The square magnifier has 80 LED lights, while the round models have 90 or 36
  • Articulated arm (800 mm) for a 200° rotation and better manipulation
  • Cover to protect the lens from dust and scratches when the magnifier is not being used
Designed for biological and clinical laboratories, electronics and chemical industries, alloy and jewellery manufacturing.
Electrical data: 270 mA, 100 to 240 V, 50 to 60 Hz, 6,5 W (square magnifiers); 300 mA, 100 to 240 V, 50 to 60 Hz, 7 W (round magnifiers)
Bestillingsoplysninger: Includes a table bracket.