VWR®, Bovine serum albumin (BSA)
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
0175-25GEA 570 DKK
0175-25G
VWR®, Bovine serum albumin (BSA)
Proteiner
High buffering capacity. High ionic strength. Permits extended electophoresis runs without buffer recirculation.
A heat shock isolation product provided as a fine, white powder in a salt-free form (pH 5,2) for low salt applications. Store cold.
Biotechnology Grade: Materials equivalent to Ultra Pure, but particularly suitable for use in Molecular Biology applications. Tested for specific contaminants such as nucleases and bacteria where appropriate.
Heavy metals: 0,005%
Loss on Drying: 2,0%
pH (2%, water) @25 °C: 4,9 to 5,3
Purity (electrophoresis): 96,0%
Solubility (2%. water): Pass