Water stills for double distillation
Leverandør: GFL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
171-3024EA 55010 DKK
GFLA2104 GFLA2102 GFLA2108
Water stills for double distillation
These fully automatic water stills produce ultra-pure, low-gas, bacteria and pyrogen free distillate with a very low conductivity.
  • Suitable for both bench and wall mounting
  • Electronic level control during the entire distilling process
  • Low energy consumption due to distillation of heated cooling water
  • Water saving through automatic water cut-off; unnecessary water consumption will be avoided
  • Condenser (cooler): 1st stage made of stainless steel; 2nd stage including the baffle made of DURAN® Borosilicate glass 3.3
  • Evaporator easily accessible for cleaning purposes
  • CO₂ degassing through outlet in the condenser
  • Water supply through a built-in solenoid valve with connection for pressure hose ¹/₂" (inner Ø approx. 12,7 mm)
  • Cooling water outlet with hose connection ³/₄" (Ø approx. 19,0 mm)
Electronic impurity detector switches the unit off in case of high degree impurities of the water in the evaporator 1st stage, the red pilot lamp 'Clean' will glow. Double-action low water cut-off through float switch and thermostatic safety switch.
The distillate is used in research and development like bacteriological and medical preparation of samples and for the preparation of cell and tissue cultures. It is also used for cleaning and sterilisation processes, for buffer solutions and for microbiological and analytical applications, especially for HPLC.