Trousers for men, C.I., Design C, dark grey
Leverandør: Fristads Kansas Group
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KWIN843941131C156EA 0 DKK
KWIN843941131C156 KWIN843941539C152 KWIN843941131C154 KWIN843941539C150 KWIN843941131C152 KWIN843941131C150 KWIN843941530C156 KWIN843941520C150 KWIN843941530C56 KWIN843941520C152 KWIN843941530C54 KWIN843941940C46 KWIN843941539C156 KWIN843941210C146 KWIN843941940C48 KWIN843941210C148 KWIN843941539C154 KWIN843941530C58 KWIN843941530C60 KWIN843941940C54 KWIN843941530C150 KWIN843941940C56 KWIN843941530C152 KWIN843941520C154 KWIN843941940C50 KWIN843941520C156 KWIN843941530C154 KWIN843941530C62 KWIN843941940C52 KWIN843941730C60 KWIN843941730C62 KWIN843941520C148 KWIN843941940C44 KWIN843941539C48 KWIN843941730C56 KWIN843941539C46 KWIN843941730C58 KWIN843941539C44 KWIN843941520C146 KWIN843941131C62 KWIN843941941C156 KWIN843941941C152 KWIN843941640C156 KWIN843941941C154 KWIN843941131C60 KWIN843941331C156 KWIN843941640C48 KWIN843941640C46 KWIN843941210C50 KWIN843941640C44 KWIN843941210C52 KWIN843941331C150 KWIN843941640C52 KWIN843941941C44 KWIN843941640C50 KWIN843941210C44 KWIN843941941C46 KWIN843941210C46 KWIN843941331C154 KWIN843941210C48 KWIN843941331C152 KWIN843941640C152 KWIN843941131C52 KWIN843941941C150 KWIN843941131C54 KWIN843941640C154 KWIN843941131C56 KWIN843941131C58 KWIN843941640C150 KWIN843941640C148 KWIN843941941C146 KWIN843941640C146 KWIN843941131C50 KWIN843941730C150 KWIN843941530C46 KWIN843941640C58 KWIN843941530C44 KWIN843941730C152 KWIN843941640C56 KWIN843941210C60 KWIN843941941C148 KWIN843941940C58 KWIN843941530C48 KWIN843941640C54 KWIN843941210C62 KWIN843941131C48 KWIN843941210C54 KWIN843941640C62 KWIN843941210C56 KWIN843941730C154 KWIN843941940C60 KWIN843941640C60 KWIN843941530C52 KWIN843941210C58 KWIN843941730C156 KWIN843941940C62 KWIN843941530C50 KWIN843941131C44 KWIN843941131C46 KWIN843941331C54 KWIN843941440C56 KWIN843941520C52 KWIN843941331C56 KWIN843941440C58 KWIN843941520C54 KWIN843941440C52 KWIN843941331C58 KWIN843941520C50 KWIN843941440C54 KWIN843941231C46 KWIN843941440C156 KWIN843941440C154 KWIN843941231C48 KWIN843941440C152 KWIN843941941C58 KWIN843941231C44 KWIN843941440C150 KWIN843941730C146 KWIN843941730C148 KWIN843941231C50 KWIN843941941C62 KWIN843941520C48 KWIN843941440C60 KWIN843941941C60 KWIN843941440C62 KWIN843941520C44 KWIN843941331C60 KWIN843941331C62 KWIN843941520C46 KWIN843941440C46 KWIN843941331C44 KWIN843941440C48 KWIN843941331C46 KWIN843941440C44 KWIN843941331C48 KWIN843941331C148 KWIN843941331C146 KWIN843941941C48 KWIN843941941C54 KWIN843941941C56 KWIN843941440C148 KWIN843941941C50 KWIN843941941C52 KWIN843941440C146 KWIN843941231C154 KWIN843941440C50 KWIN843941231C156 KWIN843941331C50 KWIN843941231C150 KWIN843941331C52 KWIN843941231C152 KWIN843941231C146 KWIN843941231C148 KWIN843941730C52 KWIN843941940C150 KWIN843941730C54 KWIN843941730C50 KWIN843941940C156 KWIN843941539C58 KWIN843941940C154 KWIN843941730C46 KWIN843941940C152 KWIN843941539C56 KWIN843941730C48 KWIN843941539C54 KWIN843941539C52 KWIN843941539C50 KWIN843941520C62 KWIN843941131C146 KWIN843941520C60 KWIN843941231C56 KWIN843941539C148 KWIN843941530C146 KWIN843941231C58 KWIN843941730C44 KWIN843941539C146 KWIN843941210C156 KWIN843941231C52 KWIN843941530C148 KWIN843941231C54 KWIN843941210C152 KWIN843941231C60 KWIN843941940C148 KWIN843941231C62 KWIN843941210C154 KWIN843941940C146 KWIN843941210C150 KWIN843941539C62 KWIN843941520C56 KWIN843941539C60 KWIN843941520C58 KWIN843941131C148
Trousers for men, C.I., Design C, dark grey
Arbejdstøj
Robust trousers made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro® closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring, inside knee pockets
Weight: 260 g/m²
Basic colour: Dark grey
Bestillingsoplysninger: Trousers are available in 12 different accent colours.