DAB substrate system for biotechnology
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E885-200MLEA 807 DKK
VWRCE885-200ML
DAB substrate system for biotechnology
Commonly used for detection of horseradish peroxidase in membrane-bound and in situ applications. The insoluble end product is resistant to alcohol and fading, making it ideal for immunohistochemistry.
Leveringsinformation: Complete substrate system includes 20 DAB substrate tablets, peroxidase solution and a mixing/dispensing dropper bottle for fast and easy preparation. Each tablet will yield 10 ml of substrate solution, enough to effectively detect 50 cm² of membrane or about 20 slides.