TRIS-borate-EDTA (TBE) buffer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J490-2PKEA 524 DKK
VWRCJ490-2PK
TRIS-borate-EDTA (TBE) buffer
TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose.
Foil pouch contains sufficient material to prepare one L of 10× concentrate.
A single strength (1×) solution contains 0.089 M Tris Base; 0,089 M Boric acid; and 0,002 M EDTA disodium salt dihydrate with a final pH of 8,3.