3-Hydroxypicolinamide ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAH0786-5GEA 1200 DKK
TCIAH0786-5G
3-Hydroxypicolinamide ≥98.0% (by titrimetric analysis)
3-Hydroxypicolinamide
Formel: C₆H₆N₂O₂
Molekylvægt: 138,12 g/mol
MDL Number: MFCD00006302
CAS-nummer: 933-90-4
EINECS: 213-274-3
https://dk.vwr.com/stibo/web/std.lang.all/08/17/28970817.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR