4-Cyano-4'-hexyloxybiphenyl ≥97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H55876.06EA
4060
DKK
H55876.06
H55876.03
4-Cyano-4'-hexyloxybiphenyl ≥97%
4-Cyano-4'-hexyloxybiphenyl
Formel:
C₁₉H₂₁NO Molekylvægt: 279,38 g/mol Smeltepunkt: 56…58 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00134145 CAS-nummer: 41424-11-7 EINECS: 255-358-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...