4-Cyano-4'-heptyloxybiphenyl ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC1606-1GEA 337 DKK
TCIAC1606-1G
4-Cyano-4'-heptyloxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-heptyloxybiphenyl
Formel: C₂₀H₂₃NO
Molekylvægt: 293,41 g/mol
MDL Number: MFCD00134146
CAS-nummer: 52364-72-4
EINECS: 257-876-4
https://dk.vwr.com/stibo/web/std.lang.all/88/23/28938823.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 52.0 to 77.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR