Choline iodide ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC2573-25GEA 591 DKK
TCIAC2573-25G
Choline iodide ≥98.0% (by titrimetric analysis)
Choline iodide
Formel: C₅H₁₄INO
Molekylvægt: 231,08 g/mol
MDL Number: MFCD00011900
CAS-nummer: 17773-10-3
EINECS: 241-754-2
https://dk.vwr.com/stibo/web/std.lang.all/95/71/28939571.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR