1-Methyl-5-(tributylstannyl)imidazole ≥90%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H51405.03EA
2480
DKK
H51405.03
1-Methyl-5-(tributylstannyl)imidazole ≥90%
1-Methyl-5-(tributylstannyl)imidazole
Formel:
C₁₆H₃₂N₂Sn Molekylvægt: 371,15 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01319031 CAS-nummer: 147716-03-8 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...