Propargyl-p-toluenesulphonate ≥96.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAT1455-5GEA
1320
DKK
TCIAT1455-5G
Propargyl-p-toluenesulphonate ≥96.0% (by GC)
Propargyl-p-toluenesulphonate
Formel:
C₁₀H₁₀O₃S Molekylvægt: 210,25 g/mol |
MDL Number:
MFCD00078365 CAS-nummer: 6165-76-0 |
Specifikationer Testresultater
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...