Safety blood lancets, VITREX® STERILANCE® Lite II
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VITREX® STERILANCE®
233-0136EA 128 DKK
MODU378113
Safety blood lancets, VITREX® STERILANCE® Lite II
Safety lancets for blood glucose and other in vitro diagnostic tests.
  • Needle fully shielded before and after use
  • Pre-loaded pressure activated design
  • High quality, ultra sharp tri-bevel needle, high speed penetration
A variety of point configurations and penetration depths suit most capillary blood requirements.