3-Propylthiophene ≥97.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAP1128-1GEA 591 DKK
TCIAP1128-1G TCIAP1128-5G
3-Propylthiophene ≥97.0% (by GC)
3-Propylthiophene
Formel: C₇H₁₀S
Kogepunkt: 161°C
MDL Number: MFCD00143183
CAS-nummer: 1518-75-8
UN nummer: 1993
ADR: 3,III
https://dk.vwr.com/stibo/web/std.lang.all/17/57/28991757.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR