3-Phenylpropionitrile ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAP0611-25GEA 370 DKK
TCIAP0611-25G
3-Phenylpropionitrile ≥98.0% (by GC)
3-Phenylpropionitrile
Formel: C₉H₉N
Molekylvægt: 131,18
Kogepunkt: 254°C
Flammepunkt: 113°C
MDL Number: MFCD00001961
CAS-nummer: 645-59-0
EINECS: 211-447-8
UN nummer: 3276
ADR: 6.1,III
https://dk.vwr.com/stibo/web/std.lang.all/13/41/28991341.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR