3-Phenylpropionitrile 99+%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

ACRO173990100EA 234 DKK
ACRO173990100
3-Phenylpropionitrile 99+%
3-Phenylpropionitrile
Formel: C₉H₉N
Molekylvægt: 131,18
Smeltepunkt: 2…1 °C
Massefylde: 1 g/cm³
Flammepunkt: >112 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001961
CAS-nummer: 645-59-0
EINECS: 211-447-8
UN nummer: 3276
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5230 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR