tert-Butyl benzoate ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAB0067-25MLEA
206
DKK
TCIAB0067-25ML
tert-Butyl benzoate ≥98.0% (by GC)
tert-Butyl benzoate
Formel:
C₁₁H₁₄O₂ Molekylvægt: 178,2277 g/mol Kogepunkt: 54°C |
MDL Number:
MFCD00048244 CAS-nummer: 774-65-2 EINECS: 212-267-2 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...