2-Methyl-5-(trifluoromethoxy)benzonitrile ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H34142.06EA 4760 DKK
H34142.06 H34142.03
2-Methyl-5-(trifluoromethoxy)benzonitrile ≥97%
2-Methyl-5-(trifluoromethoxy)benzonitrile
Formel: C₉H₆F₃NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18399750
CAS-nummer: 1261679-13-3
UN nummer: 3276
ADR: 6.1,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR