(3-Ethoxy-2,6-difluorophenyl)acetic acid ≥97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H32859.MDEA
712
DKK
H32859.MD
H32859.03
(3-Ethoxy-2,6-difluorophenyl)acetic acid ≥97%
(3-Ethoxy-2,6-difluorophenyl)acetic acid
Formel:
C₁₀H₁₀F₂O₃ Molekylvægt: 216,18 g/mol Smeltepunkt: 101…103 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11519374 CAS-nummer: 1092461-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...