4-Ethoxy-2-fluorobenzonitrile ≥97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H33730.06EA
3970
DKK
H33730.06
H33730.03
4-Ethoxy-2-fluorobenzonitrile ≥97%
4-Ethoxy-2-fluorobenzonitrile
Formel:
C₉H₈FNO Molekylvægt: 165,17 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD13196591 CAS-nummer: 186191-34-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...