3-Ethoxypropionitrile ≥99.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAE0299-25MLEA
132
DKK
TCIAE0299-25ML
TCIAE0299-500ML
3-Ethoxypropionitrile ≥99.0% (by GC)
3-Ethoxypropionitrile
Formel:
C₅H₉NO Molekylvægt: 99,1300 g/mol Kogepunkt: 171°C Flammepunkt: 63°C |
MDL Number:
MFCD00001959 CAS-nummer: 2141-62-0 EINECS: 218-393-4 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...