3-Butoxypropionitrile ≥96.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB0959-25MLEA
452
DKK
TCIAB0959-25ML
3-Butoxypropionitrile ≥96.0% (by GC)
3-Butoxypropionitrile
Formel:
C₇H₁₃NO Molekylvægt: 127,19 g/mol Kogepunkt: 206°C Flammepunkt: 89°C |
MDL Number:
MFCD00045643 CAS-nummer: 6959-71-3 EINECS: 230-153-0 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...