(4-Methoxyphenyl)magnesium bromide 1 mol/l
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H54317.AEEA
1570
DKK
H54317.AE
(4-Methoxyphenyl)magnesium bromide 1 mol/l
(4-Methoxyphenyl)magnesium bromide
Formel:
C₇H₇BrMgO Molekylvægt: 211,34 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013515 UN nummer: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...