4-(Trifluoromethoxy)thiobenzamide ≥97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H58585.03EA
2230
DKK
H58585.03
4-(Trifluoromethoxy)thiobenzamide ≥97%
4-(Trifluoromethoxy)thiobenzamide
Formel:
C₈H₆F₃NOS Molekylvægt: 221,19 g/mol Smeltepunkt: 124…126 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00068113 CAS-nummer: 149169-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...