2-Amino-4,5-dimethoxythiobenzamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H52243.MDEA 762 DKK
H52243.MD
2-Amino-4,5-dimethoxythiobenzamide 97%
2-Amino-4,5-dimethoxythiobenzamide
Formel: C₉H₁₂N₂O₂S
Molekylvægt: 212,27 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16658867
CAS-nummer: 1379207-02-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR