2-Cyanobenzamide ≥96.0% (by total nitrogen basis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC0431-1GEA
534
DKK
TCIAC0431-1G
2-Cyanobenzamide ≥96.0% (by total nitrogen basis)
2-Cyanobenzamide
Formel:
C₈H₆N₂O Molekylvægt: 146,15 g/mol |
MDL Number:
MFCD00014797 CAS-nummer: 17174-98-0 EINECS: 241-224-0 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 96.0 % |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...