4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid ≥98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H52551.03EA
4000
DKK
H52551.03
4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid ≥98%
4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid
Formel:
C₁₀H₁₁BClNO₃ Molekylvægt: 239,46 g/mol Smeltepunkt: 238…242 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07783865 CAS-nummer: 871332-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...