4-Bromo-N-(4-fluorophenyl)benzamide ≥97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H57409.03EA 5000 DKK
H57409.03 H57409.MD
4-Bromo-N-(4-fluorophenyl)benzamide ≥97%
4-Bromo-N-(4-fluorophenyl)benzamide
Formel: C₁₃H₉BrFNO
Molekylvægt: 294,12 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00442009
CAS-nummer: 346723-41-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR